Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Epitetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific

Supplier: Thermo Scientific
23312-5000 23312-1000
TS23312-1000EA 204 USD InStock
TS23312-1000 TS23312-5000
4-Epitetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific
Standards Organic Standards
CAS No.: 23313-80-6

MDL No.: MFCD00865028
GHS Symbols: GHS07, GHS08,
Melting Point: 217 °C (D)
Appearance: Yellow Crystalline powder
Storage Temperature: Freezer
Signal word: Warning

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty