Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Epioxytetracycline, 97%, can be used as secondary standard

Supplier: Thermo Scientific
25771-1000
TS25771-1000EA 257 USD InStock
TS25771-1000
4-Epioxytetracycline, 97%, can be used as secondary standard
Standards Organic Standards
CAS No.: 14206-58-7

MDL No.: MFCD00021181
GHS Symbols: GHS07, GHS08,
Melting Point: 168 °C (D)
Appearance: Green-beige to brown Powder
Storage Temperature: Ambient
Signal word: Warning

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty