Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Terpineol 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Methylcyclohex-3-en-1-yl)propan-2-olTerpineolα-Terpineol

301610025 301610250 301615000
TS301610025EA 181 USD InStock
TS301610025 TS301610250 TS301615000
(±)-α-Terpineol 97+%
(±)-α-Terpineol


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 214…224 °C (760 mmHg)
Melting Pt: 31…34 °C
Density: 0.937 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
Order Now
Specification Test Results
Appearance Clear colorless to white solid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4850 (20°C, 589 nm)
GC ≥97.0 %
Specific gravity (20°C) 0.934 to 0.940

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty