Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Aspartyl-L-Phenylalanine

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

31002-5000 31002-1000
TS31002-5000EA 254 USD InStock
TS31002-5000 TS31002-1000
L-Aspartyl-L-Phenylalanine
Proteins and Peptides
  • Infrared spectrum: Conforms
  • Appearance of solution: (5% in CH₃COOH 80%) clear colorless
  • Specific optical rotation: 10.0 to 13.0° (24 °C, 589 nm) (c=1, 0.5 N HCl)
  • Melting point: 236 to 239 °C

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty