Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform 99.9% stabilized, ECD tested for residue analysis, for pesticide residue analysis residue <0.0005%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

32667-0025
TS32667-0025EA 171 USD InStock
TS32667-0025
Chloroform 99.9% stabilized, ECD tested for residue analysis, for pesticide residue analysis residue <0.0005%
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.01 % (Coulometric)
GC ≥99.9 %
Color scale ≤10 APHA
Stabilizer 0.0035 to 0.005 %w/w Amylene
GC/ECD (as lindane) ≤5 ng/l
Residue after evaporation ≤0.0005 %
Acidity (HCl) ≤0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty