Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform, extra dry 99.9% stabilized, AcroSeal™

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

32682-0025 32682-0010 32682-1000
TS32682-0025EA 189.12 USD InStock
TS32682-0025 TS32682-0010 TS32682-1000
Chloroform, extra dry 99.9% stabilized, AcroSeal™
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.85 %
Stabilizer 50 to 150 ppm Amylene
Residue after evaporation ≤0.0005 %
Filt.over 0.2 micron PFTE Filtered

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty