Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran, extra dry 99.85% stabilized, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: OxolaneTHFCyclotetramethylene oxideTetramethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

32697-1000 32697-0010 32697-0025
TS32697-0025EA 249 USD InStock
TS32697-0025 TS32697-1000 TS32697-0010
Tetrahydrofuran, extra dry 99.85% stabilized, AcroSeal™
Tetrahydrofuran


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
GC ≥99.80 %
Stabilizer ≥200 ppm BHT
Residue after evaporation ≤0.001 %
Filt.over 0.2 micron PFTE Filtered
Acidity (CH3COOH) ≤0.0005 meq/g
Peroxides ≤0.0025 % (as H2O2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty