Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0

Supplier: Thermo Scientific
32734-5000
TS32734-5000EA 133.71 USD InStock
TS32734-5000
Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0
Buffers Biological Buffers
CAS No.: 38641-82-6

MDL No.: MFCD00236359
Appearance: Clear colourless solution
Storage temperature: Ambient

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty