Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform, extra dry over Molecular Sieve 99.9% stabilized, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

36432-1000
TS36432-1000EA 53 USD InStock
TS36432-1000
Chloroform, extra dry over Molecular Sieve 99.9% stabilized, AcroSeal™
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.85 %
Stabilizer 50 to 150 ppm Amylene

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty