Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

36439-0010 36439-5000
TS36439-5000EA 57.16 USD InStock
TS36439-5000 TS36439-0010
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal™
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.8 %
Acidity/alkalinity ≤0.0005 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty