Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-(4-hydroxyphenyl)propionate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(4-Hydroxy-phenyl)-propionic acid-OMeMethyl 3-(4-hydroxyphenyl)propionate3-(4-Hydroxyphenyl)propionic Acid Methyl Ester

36943-0050
TS36943-0050EA 62 USD InStock
TS36943-0050
Methyl-3-(4-hydroxyphenyl)propionate 98%
Methyl-3-(4-hydroxyphenyl)propionate
Appearance: Transparent white Low melting mass


Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 108 °C (11 mmHg)
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00071577
CAS Number: 5597-50-2
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals or fused mass
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty