Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus pentoxide - methanesulfonic acid

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Eaton's Reagent

36980-0025 36980-5000
TS36980-5000EA 171.2 USD InStock
TS36980-5000 TS36980-0025
Phosphorus pentoxide - methanesulfonic acid
Phosphorus pentoxide - methanesulfonic acid


MW: 141.95 g/mol
Boiling Pt: 122 °C (1 mmHg)
Density: 1.5 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00070545
CAS Number: 39394-84-8
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance to light brown or pale pink Solution (May contain precipitate, re-dissolves on warming)
Refractive index 1.4325 to 1.4375 (20°C, 589 nm)
Titration with NaOH 6.5 to 10.0 % (P2O5)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty