Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluidine 99%, crystalline molten mass

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Toluidine4-Aminotoluene4-Methylaniline

37635-2500 37635-0050 37635-0250 37635-0010
TS37635-0250EA 842 USD InStock
TS37635-0250 TS37635-0010 TS37635-0050 TS37635-2500
p-Toluidine 99%, crystalline molten mass
p-Toluidine


Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: 41…46 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007906
CAS Number: 106-49-0
EINECS: 203-403-1
UN: 3451
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Colorless to red Liquid after melting or solid (May discolor to brown on storage)
Infrared spectrum Conforms
Melting point 41 to 46 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty