Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide octahydrate 99.95%

Supplier: Thermo Scientific Chemicals
Danger

AC37782-0250 AC37782-0050
TS37782-0250EA 168 USD InStock
TS37782-0250 TS37782-0050
Barium hydroxide octahydrate 99.95%
Barium hydroxide octahydrate


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals or flakes and/or chunks
Total trace metal impurities ≤0.05 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty