Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, micropearls for analysis

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

38304-0025 383042500 383040010
TS38304-0025EA 177 USD InStock
TS38304-0025 TS383040010 TS383042500
Sodium hydroxide, micropearls for analysis
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Order Now
Specification Test Results
Appearance White Pearls
Titration with HCl ≥99.0 %
Sodium carbonate (Na2CO3) ≤0.4 %
Sodium sulfate (Na2SO4) ≤80 ppm
Sodium chloride (NaCl) ≤200 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤0.5 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty