Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide 99.8+% for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

39067-0010 39067-0025
TS39067-0010EA 153 USD InStock
TS39067-0010 TS39067-0025
N,N-Dimethylacetamide 99.8+% for HPLC
N,N-Dimethylacetamide


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 164…166 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.937 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.80 %
UV 1 cm cell vs HPLC-grade water
UV at 275 nm A: ≤0.700
UV at 285 nm A: ≤0.301
UV at 310 nm A: ≤0.046
Non-volatile matter ≤0.0003 %
Acidity ≤0.0005 meq/g
Alkalinity ≤0.003 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty