Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

39635-1000 AC39635-0010 39635-0025
TS39635-0025EA 266 USD InStock
TS39635-0025 TS39635-1000 TS39635-0010
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal™
N,N-Dimethylacetamide


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 164…166 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.937 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %
Residue after evaporation ≤0.005 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty