Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether, anhydrous 99+% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

AC41006-0025 AC41006-5000
TS41006-0025EA 160 USD InStock
TS41006-0025 TS41006-5000
Diethyl ether, anhydrous 99+% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.
Diethyl ether


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 34.6 °C (760 mmHg)
Melting Pt: –116 °C
Density: 0.714 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.03 % (Coulometric)
Boiling point 34 to 35 °C
Density (20/20 °C) 0.713 to 0.715
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer ≤10 ppm BHT
Residue after evaporation ≤0.001 %
Carbonyl (as HCHO) ≤0.001 %
Titrable acid ≤0.0002 meq/g
Ethanol (limit about 0.05%) Passes test
Peroxides ≤1 ppm (as H2O2)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty