Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid ≥85%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

41296-5000
TS41296-5000EA 69.76 USD InStock
TS41296-5000
Lactic acid ≥85%
Lactic acid


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Boiling Pt: 122 °C (20 hPa)
Melting Pt: 16.8 °C
Density: 1.357 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 209-954-4
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Residue after ignition ≤0.02 %
Titration excess NaOH 85.0 to 90.0 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty