Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Papain

Supplier: Thermo Scientific
41676-0100 41676-5000
TS41676-0100EA 67.95 USD InStock
TS41676-0100 TS41676-5000
Papain
Enzymes
CAS No.: 9001-73-4

MDL No.: MFCD00131791
GHS Symbols: GHS08
Appearance: Cream to light brown Powder
Storage Temperature: Refrigerator
Signal word: Danger

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty