Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Colophony (Rosin gum)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Rosin, gumGum rosinRosin

41917-0025 41917-5000
TS41917-5000EA 70 USD InStock
TS41917-5000 TS41917-0025
Colophony (Rosin gum)
Colophony (Rosin gum)


Melting Pt: 75…81 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00132205
CAS Number: 8050-09-7
EINECS: 232-475-7
Order Now
Specification Test Results
Appearance Almost white to yellow to orange Powder or flakes or chunks
Infrared spectrum Conforms
Melting point 72 to 83 °C
Acid value 156 to 182 mg KOH/g
Uniformity coefficient ≤1.8
Moisture holding capacity 53 to 58 % (H+)
Total volume capacity ≥1.8 eq/l (H+)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty