Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide octahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

AC42346-5000 AC42346-0025 42346-1000
TS42346-5000EA 129.43 USD InStock
TS42346-5000 TS42346-0025 TS42346-1000
Barium hydroxide octahydrate 98+% ACS
Barium hydroxide octahydrate


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Boiling Pt: 780 °C (1013 hPa)
Melting Pt: 78 °C
Density: 2.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals or flakes
Titration with HCl ≥98.0 %
Chloride (Cl) ≤0.001 %
Sulfide (S) (Limit about 0.001 %) Passes test
Carbonate (as BaCO3) ≤2.0 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Strontium (Sr) ≤0.8 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 % (in dilute HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty