Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid ∼37% in water ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Muriatic acidHydrochloric acid solution

42379-0026 42379-5001 423790251 42379-5000 42379-0025
TS42379-5000EA 52.22 USD InStock
TS42379-5000 TS42379-5001 TS42379-0025 TS42379-0026 TS423790251
Hydrochloric acid ∼37% in water ACS
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear solution
Color scale ≤10 APHA
Residue after ignition ≤0.0005 %
Titration with NaOH 36.5 to 38.0 %
Bromide (Br) ≤50 ppm
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Free chlorine ≤1 ppm (as Cl)
Ammonium (NH4) ≤3 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (ICP-OES) ≤1 ppm
Extractable organic subst (about 5 ppm) Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty