Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitromethane 96% ACS

Supplier: Thermo Scientific Chemicals
Warning

42401-5000 42401-0025 42401-0010
TS42401-5000EA 111.62 USD InStock
TS42401-5000 TS42401-0025 TS42401-0010
Nitromethane 96% ACS
Nitromethane


Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 101.2 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.12 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Merck Index: 12,06708
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3805 to 1.3825 (20°C, 589 nm)
Water ≤0.05 % (Coulometric)
GC ≥95 %
Color scale ≤10 APHA

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty