Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 97+%, pellets ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

42433-0251 42433-5000 42433-0100 424330010 42433-0025 424330250
TS42433-0025EA 179.09 USD InStock
TS42433-0025 TS424330250 TS424330010 TS42433-5000 TS42433-0100 TS42433-0251
Sodium hydroxide 97+%, pellets ACS
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Order Now
Specification Test Results
Appearance White Pellets
Titration with HCl ≥97.0 %
Chloride (Cl) ≤50 ppm
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Sodium carbonate (Na2CO3) ≤1.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤10 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Ag) ≤20 ppm
Nitrogen compounds (as N) ≤0.001 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty