Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionaldehyde 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propanal

42721-1000
TS42721-1000EA 51.64 USD InStock
TS42721-1000
Propionaldehyde 99+%, AcroSeal™
Propionaldehyde


Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –81 °C
Density: 0.798 g/cm³
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
UN: 1275
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA
Acidity/alkalinity ≤0.00008 meq/g
Substance darkened by H2SO4 passes test
Calcium (Ca) ≤0.2 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Hydrocarbons ≤0.1 % Aromatic hydrocarbons
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤10 ppm
Bromine number ≤0.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty