Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane 95%, Technical Grade

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Pentane

43403-0250 AC43403-0050
TS43403-0050EA 235 USD InStock
TS43403-0050 TS43403-0250
n-Pentane 95%, Technical Grade
n-Pentane


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –130 °C
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3570 to 1.3590 (20°C, 589 nm)
GC ≥94 %
UV at 265 nm A: ≤1.0
UV at 270 nm A: ≤0.523
UV at 280 nm A: ≤0.301
UV at 290 nm A: ≤0.155
UV at 310 nm A: ≤0.097
UV at 330 nm A: ≤0.046
UV at 400 nm A: ≤0.009
Residue after evaporation ≤0.001 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty