Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium dihydrogen phosphate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Ammonium dihydrogenphosphateAmmonium phosphate, monobasicAmmonium Phosphate Monobasic

AC43679-5000 43679-1000 AC43679-0250
TS43679-5000EA 532 USD InStock
TS43679-5000 TS43679-0250 TS43679-1000
Ammonium dihydrogen phosphate 99.999% (trace metals basis)
Ammonium dihydrogen phosphate


Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
Order Now
Specification Test Results
Appearance Colorless to white Crystals
Total trace metal impurities ≤10 ppm
Titration with NaOH ≥97.5 %

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty