Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 30% in water, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

44435-0050
TS44435-0050EA 61.95 USD InStock
TS44435-0050
Sodium hydroxide 30% in water, Technical Grade
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 120 °C (760 mmHg)
Melting Pt: 8 °C
Density: 1.31 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration with HCl 28.0 to 33.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty