Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate dihydrate ACS

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateTrisodium citrateSodium citrate dihydrateCitric acid tridosium salt dihydrate

AC44633-2500 AC44633-0050 AC44633-0010
TS44633-0010EA 70 USD InStock
TS44633-0010 TS44633-0050 TS44633-2500
tri-Sodium citrate dihydrate ACS
tri-Sodium citrate dihydrate


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
pH 7.0 to 9.0 (5 % in water at 25.0°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.003 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Ammonia (NH3) ≤0.003 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty