Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',7'-Dichlorofluorescein ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

44693-0010 44693-0100
TS44693-0100EA 199.38 USD InStock
TS44693-0100 TS44693-0010
2',7'-Dichlorofluorescein ACS
2',7'-Dichlorofluorescein


Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Melting Pt: 280 °C (dec.)
Density: 0.79 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS Number: 76-54-0
Order Now
Specification Test Results
Appearance Light orange to red-brown Powder
Suitability (as adsorption indicator) Passes test
Clarity of alcohol solution Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty