Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium dihydrogen phosphate 99% for HPLC

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Ammonium dihydrogenphosphateAmmonium phosphate, monobasicAmmonium Phosphate Monobasic

44738-2500
TS44738-2500EA 257.93 USD InStock
TS44738-2500
Ammonium dihydrogen phosphate 99% for HPLC
Ammonium dihydrogen phosphate


Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Water ≤1.0 %
Solubility (10 % in water) Clear colorless
UV (10 % in water)
UV at 260 nm A: ≤0.05
UV at 280 nm A: ≤0.05
pH 4.0 to 5.0 (10 % in water)
Titration with NaOH ≥98.5 %

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty