Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium chloride hexahydrate 98+%, Extra Pure

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


AC44770-0010 AC44770-0050
TS44770-0050EA 344 USD InStock
TS44770-0050 TS44770-0010
Magnesium chloride hexahydrate 98+%, Extra Pure
Magnesium chloride hexahydrate


Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Melting Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Complexometric 98.0 to 101.0 %
Water 51.0 to 55.0 %
Heavy metals (as Pb) ≤10 ppm

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty