Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium dihydrogen phosphate 98+% for biochemistry

Supplier: Thermo Scientific Chemicals
Synonyms: Ammonium dihydrogenphosphateAmmonium phosphate, monobasicAmmonium Phosphate Monobasic

AC44784-2500 AC44784-0010
TS44784-2500EA 53 USD InStock
TS44784-2500 TS44784-0010
Ammonium dihydrogen phosphate 98+% for biochemistry
Ammonium dihydrogen phosphate


Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
Order Now
Specification Test Results
Appearance Colorless to white Crystalline powder or crystals
UV (0.1 M in water)
UV at 260 nm A: ≤0.06
UV at 280 nm A: ≤0.05
pH 3.8 to 4.5 (5 % in water)
Titration with NaOH ≥98.0 %
Chloride (Cl) ≤0.0005 %
Nitrate (NO3) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty