Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 98%, flakes, pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

450580250 450580100 45058-0025
TS450580100EA 194 USD InStock
TS450580100 TS45058-0025 TS450580250
Sodium hydroxide 98%, flakes, pure
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Appearance White Flakes
Titration with HCl ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty