Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Alanyl-L-Glutamine

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

45161-0010
TS45161-0010EA 45 USD InStock
TS45161-0010
L-Alanyl-L-Glutamine
Proteins and Peptides
  • Infrared spectrum: Conforms
  • Loss on drying: ≤1%
  • Residue after ignition: ≤0.1%
  • Specific optical rotation: 8.0 to 12.0° (20 °C, 589 nm) (C=5, H₂O)
  • Heavy metals (as Pb): ≤10 ppm
  • Melting point: 213 to 220 °C

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty