Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethylamine for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: TEA(Diethylamino)ethane

46081-2500
TS46081-2500EA 83.78 USD InStock
TS46081-2500
Triethylamine for HPLC
Triethylamine


Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.728 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
UN: 1296
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/82/729282.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥99.4 %
UV at 254 nm A: ≤0.125 (0.1 M in water)
Residue after evaporation ≤0.01 %
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.02 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty