Would you like to log in again?
Tergitol™ 15-S-9, Non-Ionic Sufactant, Thermo Scientific
Supplier: Thermo Scientific
Tergitol™
AC46425-2500 AC46425-0100 AC46425-0010 AC46425-0025
TS46425-0010EA 81.19 USD InStock
TS46425-0010 TS46425-0100 TS46425-0025 TS46425-2500
Tergitol™ 15-S-9, Non-Ionic Sufactant, Thermo Scientific
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ 15-S-9 is a linear non-ionic surfactant that is a secondary ethoxylated alcohol.
  • Readily biodegradable
  • Excellent wetting ability
  • non-APE based
Density: 1.0100 g/ml
Caution: TERGITOL is a trademark of The Dow Chemical Company
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty