Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Casamino Acids, Powder, Thermo Scientific

Supplier: Thermo Scientific
61204-5000 61204-1000
TS61204-5000EA 261.88 USD InStock
TS61204-5000 TS61204-1000
Casamino Acids, Powder, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
MDL No.: MFCD00217199
Storage Temperature: Ambient

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty