Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether, anhydrous ACS

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

61508-5000 61508-0040 61508-0010
TS61508-5000EA 83.7 USD InStock
TS61508-5000 TS61508-0040 TS61508-0010
Diethyl ether, anhydrous ACS
Diethyl ether


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –116 °C
Density: 0.714 g/cm³ (20 °C)
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
UN: 1155
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Assay ≥99.0 %
Water ≤0.01 %
Density (25 °C) ≤0.7079
Additional info Alcohol: Passes test (limit is about 0.05%)
Additional info BHT: report (About 7.5 ppm)
Color scale ≤10 APHA
Identification passes test
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 passes test
Copper (Cu) ≤0.1 ppm
Nickel (Ni) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Carbonyl (as HCHO) ≤0.001 %
Titrable acid ≤0.0002 meq/g
Peroxides ≤1 ppm (as H2O2)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty