Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol ≥95% ACS for spectrophotometry (190 Proof)

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ethyl alcohol

61511-0010 61511-0040
TS61511-0010EA 179.89 USD InStock
TS61511-0010 TS61511-0040
Ethanol ≥95% ACS for spectrophotometry (190 Proof)
Ethanol


Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78 °C
Flash Pt: 17 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3605 to 1.3625 (25 °C, 589 nm)
Assay ≥95.0 % v/v
Solubility (in water) Passes test
UV at 210 nm A: ≤0.40
UV at 220 nm A: ≤0.25
UV at 230 nm A: ≤0.15
UV at 240 nm A: ≤0.05
UV at 270 nm A: ≤0.01
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Titrable acid ≤0.0005 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty