Would you like to log in again?
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6A20ST1T 6A20ST1 6A20ST1F
TS6A20ST1PK 86.27 USD InStock
TS6A20ST1 TS6A20ST1F TS6A20ST1T
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Septa
Use Thermo Scientific™ SureSTART™ 20 mm stoppers with crimp vials that have a 20 mm opening and a secure push fit, including our SureSTART 20 mm crimp headspace vials (level 2).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty