Would you like to log in again?
SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Supplier: Thermo Fisher Scientific
SureSTART™
6A20ST20 6A24ST26
TS6A24ST26PK 123.36 USD InStock
TS6A24ST26 TS6A20ST20
SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Septa
Level 2 septa are manufactured and tested to ensure low bleeding, robustness, and reproducibility of results for your routine GC-MS and LC-MS analyses.
  • Autoclavable
  • Single use
  • Non sterile
Choose all-purpose silicone/PTFE septa manufactured with a shore value of 50 to reduce autosampler needle issues.
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty