Would you like to log in again?
SureSTART™ Caps for 2 ml Glass Crimp Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6ACC11ST1 6ACC11NR1B 6ACC11ST1T 6ACC11NR1G 6ACC11RT1F 6ACC11RT1T 6ACC11NR1 6ACC11RT1 6ACC11ST1F 6ACC11NR1R
TS6ACC11ST1EA 36.46 USD InStock
TS6ACC11ST1 TS6ACC11RT1 TS6ACC11RT1F TS6ACC11NR1 TS6ACC11ST1F TS6ACC11NR1G TS6ACC11RT1T TS6ACC11ST1T TS6ACC11NR1B TS6ACC11NR1R
SureSTART™ Caps for 2 ml Glass Crimp Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Closures Chromatography Vial Closures
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
Use these vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. They have very low metal content, extractables, and leachables and conform to high standards so that you can meet your regulatory requirements. They are specification certified to ensure their physical dimensions and autosampler compatibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty