Would you like to log in again?
SureSTART™ Glass Crimp Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6ACV11-1PF 6ACV11-1P 6ACV11-2P 6ACV11-1PT 6ACV11-2PF 6ACV11-2PT
TS6ACV11-2PFCS 1119.76 USD InStock
TS6ACV11-2PF TS6ACV11-2PT TS6ACV11-1PF TS6ACV11-1P TS6ACV11-2P TS6ACV11-1PT
SureSTART™ Glass Crimp Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
  • Available in clear and amber glass
  • Meet all requirements of US, EU, JPN Pharmacopeia
  • Use 11 mm crimp caps and septa to seal these vials
  • Use manual or electronic crimpers and decrimpers to seal and remove the crimp caps
  • Buy in bulk for greater savings
Use these vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. They have very low metal content, extractables and leachables and conform to high standards so that you can meet your regulatory requirements. They are specification certified to ensure their physical dimensions and autosampler compatibility.
Performance Level 2 vials – for high-throughput applications:
- Identified by two waves on the vial
- Convenient write-on patch
- Suitable for GC, HPLC/UHPLC, and mass spectrometry (MS) applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty