Would you like to log in again?
SureSTART™ Glass Crimp Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6ACV20-1PEF 6ACV20-1RF 6ACV20-2PEF 6ACV20-2PE 6ACV20-1RT 6ACV20-1PE 6ACV20-1R 6ACV20-1PET 6ACV20-2PET
TS6ACV20-1RTCS 415.35 USD InStock
TS6ACV20-1RT TS6ACV20-1RF TS6ACV20-1PE TS6ACV20-2PE TS6ACV20-2PET TS6ACV20-1PEF TS6ACV20-1PET TS6ACV20-2PEF TS6ACV20-1R
SureSTART™ Glass Crimp Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ 20 ml glass crimp top headspace vials in your high-throughput volatile gas analyzes. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems.
These headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyzes. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure their physical dimensions and autosampler compatibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty