Would you like to log in again?
SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications
Supplier: Thermo Scientific
SureSTART™
6AK599W 6AK180W 6AK192W 6AK186W 6AK592W 6AK94W 6AK80W 6AK86W 6AK594W 6AK196W 6AK93BW 6AK195W 6AK95W 6AK82W 6AK188W 6AK92W 6AK78BW
TS6AK186WPK 92.5 USD InStock
TS6AK186W TS6AK592W TS6AK195W TS6AK594W TS6AK196W TS6AK180W TS6AK192W TS6AK599W TS6AK188W TS6AK82W TS6AK92W TS6AK80W TS6AK78BW TS6AK86W TS6AK93BW TS6AK95W TS6AK94W
SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications
Vials
Meet your certification and documentation needs with our level 2 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits. Select level 2 products for high throughput applications, when robustness and reproducibility are key.
  • Pre-slit and bonded septa options are available
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals, consistent data, and increased sample throughput.
All SureSTART™ vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.
SureSTART™ vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.
Level 1: Everyday analysis
Chromatography approved SureSTART™ Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
Level 2: High-throughput analysis
When robustness and reproducibility are key SureSTART™ Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
Level 3: High performance
When sensitivity is a must SureSTART™ Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty