Would you like to log in again?
SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications
Supplier: Thermo Scientific
SureSTART™
6AK599W 6AK180W 6AK192W 6AK186W 6AK592W 6AK94W 6AK80W 6AK86W 6AK594W 6AK196W 6AK93BW 6AK195W 6AK95W 6AK82W 6AK188W 6AK92W 6AK78BW
TS6AK186WPK 92.5 USD InStock
TS6AK186W TS6AK592W TS6AK195W TS6AK594W TS6AK196W TS6AK180W TS6AK192W TS6AK599W TS6AK188W TS6AK82W TS6AK92W TS6AK80W TS6AK78BW TS6AK86W TS6AK93BW TS6AK95W TS6AK94W
SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications
Vials
Meet your certification and documentation needs with our level 2 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits. Select level 2 products for high throughput applications, when robustness and reproducibility are key.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals, consistent data, and increased sample throughput.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty