Would you like to log in again?
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6APRCC20-ST1F 6ACC20-PFT 6APRCC20-ST1 6AMCC20-ST3T 6AMCC20-STH 6APRCC20-ST1T 6AMCC20-STHF 6ACC20-ST1 6AMCC20-ST3F 6AMCC20-ST3 6ACC20-ST1T 6ACC20-PFTF 6ACC20-ST1F 6AMCC20-STHT 6ACC20-PFTT
TS6ACC20-ST1EA 83.17 USD InStock
TS6ACC20-ST1 TS6AMCC20-STHT TS6ACC20-ST1F TS6APRCC20-ST1 TS6AMCC20-ST3 TS6ACC20-PFTT TS6AMCC20-STHF TS6ACC20-PFT TS6AMCC20-ST3T TS6APRCC20ST1T TS6AMCC20-STH TS6APRCC20ST1F TS6ACC20-ST1T TS6AMCC20-ST3F TS6ACC20-PFTF
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 20 mm crimp caps with crimp vials that have a 20 mm opening, including our SureSTART 10 ml and 20 ml glass crimp top headspace vials (level 2). Select level 2 products for high-throughput applications, when robustness and reproducibility are key.
Choose from aluminum and magnetic steel caps with silicone/PTFE or butyl/PTFE septa of various hardness values designed to reduce autosampler needle issues. Caps with pressure release technology will split to allow the safe release of high internal vial pressures. Use Thermo Scientific™ manual or electronic crimpers and decappers for easy application and removal of crimp caps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty