Would you like to log in again?
SureSTART™ Screw Caps, 9 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SureSTART™
6ASC9ST1R 6ASC9STB1 6ASC9RT1RT 6ASC9STBS1 6ASC9STBS1F 6ASC9STBS1T 6ASC9RT1T 6ASC9ST1F 6ASC9STB1F 6ASC9ST1RT 6ASC9ST1T 6ASC9ST1 6ASC9ST1RF 6ASC9RT1F 6ASC9RT1 6ASC9RT1R 6ASC9RT1RF 6ASC9STB1T
TS6ASC9RT1RFCS 1546.32 USD InStock
TS6ASC9RT1RF TS6ASC9STBS1T TS6ASC9ST1RF TS6ASC9STB1T TS6ASC9STBS1 TS6ASC9ST1F TS6ASC9RT1F TS6ASC9RT1RT TS6ASC9STB1 TS6ASC9STB1F TS6ASC9ST1T TS6ASC9ST1 TS6ASC9ST1R TS6ASC9STBS1F TS6ASC9RT1 TS6ASC9RT1R TS6ASC9RT1T TS6ASC9ST1RT
SureSTART™ Screw Caps, 9 mm, Level 2 High-throughput Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 9 mm screw caps with screw vials that have a 9 mm opening, including our SureSTART 2 ml glass screw vials and 1.7 ml glass high recovery screw microvials for <2 ml samples (level 2). Select level 2 products for high-throughput applications, when robustness and reproducibility are key.
Choose from polypropylene caps with all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Our AVCS™ caps incorporate advanced vial closure system technology to ensure optimal seal compression when closing a vial. Use AVCS screw caps with our SureStop™ vials for effective seals, consistent data, and increased sample throughput.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty